The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109764.1 | NP_001103234.1 | 15 - 16 | 2362 | 69 |
NM_009819.2 | NP_033949.2 | 14 - 15 | 2260 | 69 | |
NM_145732.2 | NP_663785.2 | 15 - 16 | 2362 | 69 | |
GenBank mRNA | AK158916.1 | - | 15 - 16 | 2395 | 69 |
BC051995.1 | - | 14 - 15 | 2141 | 69 | |
BC079648.1 | - | 15 - 16 | 2276 | 69 |