The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_194051.3 | NP_918940.1 | 2 - 3 | 244 | 61 |
NM_194054.3 | NP_918943.1 | 2 - 3 | 851 | 61 | |
GenBank mRNA | AK220511.1 | - | 2 - 3 | 811 | 61 |
AY102280.1 | - | 2 - 3 | 244 | 61 | |
AY102284.1 | - | 2 - 3 | 808 | 61 | |
AY114152.1 | - | 2 - 3 | 806 | 61 | |
BC056373.1 | - | 2 - 3 | 679 | 61 |