Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Urgcp
Entrez Gene ID
72046
Gene Name
upregulator of cell proliferation
Gene Aliases
2010005J08Rik, 9130001I21Rik, AW060359, Urg4, mKIAA1507
Chromosome Location
Chr.11: 5713417 - 5762436 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK041953.1 - 3 - 4 773 121

View Full Product Details