Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Purb
Entrez Gene ID
19291
Gene Name
purine rich element binding protein B
Gene Aliases
2310015K15Rik, AA114818, Cager-2, D11Bwg0414e
Chromosome Location
Chr.11: 6467599 - 6476076 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011221.3 NP_035351.1 1 - 1 1188 92
GenBank mRNA AF017630.1 - 1 - 1 1032 92
AK049443.1 - 1 - 1 325 92
AK139376.1 - 1 - 1 327 92

View Full Product Details