Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Grrp1
Entrez Gene ID
72690
Gene Name
glycine/arginine rich protein 1
Gene Aliases
2810043G13Rik, Fam110d
Chromosome Location
Chr.4: 134251102 - 134266197 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099296.2 NP_001092766.1 1 - 2 79 62
GenBank mRNA BC048704.1 - 1 - 2 75 62

View Full Product Details