Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sfxn2
Entrez Gene ID
94279
Gene Name
sideroflexin 2
Gene Aliases
C79732, F630107H02Rik
Chromosome Location
Chr.19: 46573322 - 46596901 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053196.3 NP_444426.3 6 - 7 702 78
GenBank mRNA AF325261.1 - 6 - 7 660 78
AK150722.1 - 6 - 7 653 78
BC019808.1 - 6 - 7 700 78

View Full Product Details