The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081067.1 | NP_001074536.1 | 6 - 7 | 1266 | 92 |
GenBank mRNA | BC080745.1 | - | - | 1024 | 92 |
DQ520794.1 | - | 6 - 7 | 1262 | 92 |