The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164639.1 | NP_001158111.1 | 13 - 14 | 3086 | 72 |
NM_009289.3 | NP_033315.2 | 14 - 15 | 3179 | 72 | |
GenBank mRNA | AF039574.1 | - | 14 - 15 | 3184 | 72 |
AF112855.1 | - | 13 - 14 | 2975 | 72 | |
AK037798.1 | - | 5 - 6 | 1060 | 72 | |
AK122218.1 | - | 13 - 14 | 3427 | 72 | |
BC131674.1 | - | 13 - 14 | 2987 | 72 | |
BC131675.1 | - | 14 - 15 | 3080 | 72 | |
CA535951.1 | - | 4 - 5 | 544 | 72 |