The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026831.1 | NP_081107.1 | 8 - 9 | 1186 | 90 |
GenBank mRNA | AK004148.1 | - | 8 - 9 | 1187 | 90 |
BC089625.1 | - | 8 - 9 | 1174 | 90 | |
BC132636.1 | - | 8 - 9 | 1175 | 90 | |
BC132638.1 | - | 8 - 9 | 1175 | 90 |