The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024699.1 | NP_001019870.1 | 2 - 3 | 1423 | 78 |
GenBank mRNA | AK154877.1 | - | 2 - 3 | 1501 | 78 |
AK155880.1 | - | 2 - 3 | 78 | ||
AK166382.1 | - | - | 1492 | 78 | |
BC094359.1 | - | 2 - 3 | 1423 | 78 |