Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hnrnpd
Entrez Gene ID
11991
Gene Name
heterogeneous nuclear ribonucleoprotein D
Gene Aliases
Auf1, Hnrpd
Chromosome Location
Chr.5: 99955935 - 99979015 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077265.2 NP_001070733.1 4 - 5 934 92
NM_001077266.2 NP_001070734.1 3 - 4 877 92
NM_001077267.2 NP_001070735.1 3 - 4 877 92
NM_007516.3 NP_031542.2 4 - 5 934 92
GenBank mRNA AK077409.1 - 4 - 5 922 92
BC011172.1 - 4 - 5 609 92
BC049098.1 - 4 - 5 934 92
CA451481.1 - 2 - 3 215 92
U11273.1 - 3 - 4 714 92

View Full Product Details