Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Grifin
Entrez Gene ID
77998
Gene Name
galectin-related inter-fiber protein
Gene Aliases
E130101O17Rik
Chromosome Location
Chr.5: 140563117 - 140567628 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030022.1 NP_084298.1 1 - 2 31 102
GenBank mRNA AK021365.1 - 1 - 2 32 102
BC107323.2 - 1 - 2 31 102
BC107324.1 - 1 - 2 31 102
BC107343.2 - 4 - 5 102

View Full Product Details