Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Med25
Entrez Gene ID
75613
Gene Name
mediator complex subunit 25
Gene Aliases
2610034E13Rik, 2610529E18Rik, ESTM2
Chromosome Location
Chr.7: 44876765 - 44892664 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029365.2 NP_083641.1 5 - 6 523 78
GenBank mRNA AK089940.1 - 5 - 6 524 78
AK090026.1 - 5 - 6 549 78
AK149867.1 - 6 - 7 795 78
BC021333.1 - 5 - 6 558 78
BC031138.1 - 5 - 6 566 78
BC071206.1 - 4 - 5 600 78

View Full Product Details