The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001015876.2 | NP_001015876.1 | 14 - 15 | 1930 | 61 |
GenBank mRNA | AK082884.1 | - | 14 - 15 | 61 | |
BC027065.1 | - | 9 - 10 | 1004 | 61 | |
BC039634.1 | - | 3 - 4 | 354 | 61 | |
BC068126.1 | - | 14 - 15 | 1838 | 61 |