The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145820.3 | NP_665819.2 | 13 - 14 | 2647 | 84 |
GenBank mRNA | AB085187.1 | - | 13 - 14 | 2629 | 84 |
AB085189.1 | - | 5 - 6 | 692 | 84 | |
AK013398.1 | - | 5 - 6 | 707 | 84 | |
BC075649.1 | - | 13 - 14 | 2647 | 84 | |
BC094339.1 | - | 5 - 6 | 660 | 84 | |
BC128271.1 | - | 12 - 13 | 2354 | 84 |