Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Scamp3
Entrez Gene ID
24045
Gene Name
secretory carrier membrane protein 3
Gene Aliases
Sc3, TU52
Chromosome Location
Chr.3: 89177485 - 89182770 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011886.2 NP_036016.2 8 - 9 1011 59
GenBank mRNA AF005036.1 - 8 - 9 918 59
AF295403.1 - 8 - 9 916 59
AK135335.1 - 8 - 9 995 59
AK146204.1 - 8 - 9 1013 59
AK160549.1 - 8 - 9 996 59
AK170115.1 - 7 - 8 876 59
BC002021.1 - 8 - 9 1009 59

View Full Product Details