Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tlr2
Entrez Gene ID
24088
Gene Name
toll-like receptor 2
Gene Aliases
Ly105
Chromosome Location
Chr.3: 83836272 - 83841825 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011905.3 NP_036035.3 1 - 2 89 58
GenBank mRNA AF185284.1 - 1 - 2 43 58
AF216289.1 - 1 - 2 52 58
AK005043.1 - 1 - 2 90 58
AK154504.1 - 1 - 2 59 58
BC014693.1 - 1 - 2 248 58

View Full Product Details