Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Arfrp1
Entrez Gene ID
76688
Gene Name
ADP-ribosylation factor related protein 1
Gene Aliases
1500006I01Rik, AI480700
Chromosome Location
Chr.2: 181357690 - 181365404 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001165991.1 NP_001159463.1 7 - 8 724 85
NM_001165992.1 NP_001159464.1 7 - 8 705 85
NM_001165995.1 NP_001159467.1 6 - 7 611 85
NM_029702.4 NP_083978.3 7 - 8 718 85
NR_028585.1 6 - 7 647 85
GenBank mRNA AJ413953.1 - 7 - 8 671 85
AK005174.1 - 7 - 8 666 85
AK032592.1 - 1 - 2 502 85
AK044759.1 - 7 - 8 676 85
AK050136.1 - 4 - 5 1576 85
BC010713.1 - 7 - 8 665 85
BC046782.1 - 6 - 7 585 85
BC046786.1 - 6 - 7 585 85

View Full Product Details