The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163401.1 | NP_001156873.1 | 1 - 2 | 113 | 134 |
GenBank mRNA | BY777524.1 | - | 1 - 2 | 113 | 134 |