Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrc69
Entrez Gene ID
73314
Gene Name
leucine rich repeat containing 69
Gene Aliases
1700034K16Rik
Chromosome Location
Chr.4: 14623622 - 14801320 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028499.2 NP_082775.1 3 - 4 390 107
GenBank mRNA AK006601.1 - 3 - 4 401 107
AK161371.1 - 3 - 4 393 107

View Full Product Details