The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_033527.1 | 1 - 2 | 78 | 78 | |
GenBank mRNA | AK043043.1 | - | 1 - 2 | 62 | 78 |
AK132700.1 | - | 1 - 2 | 78 | 78 | |
AK154211.1 | - | 1 - 2 | 59 | 78 |