The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113362.1 | NP_001106833.1 | 7 - 8 | 1406 | 68 |
NM_001113364.1 | NP_001106835.1 | 4 - 5 | 520 | 68 | |
NM_133910.3 | NP_598671.3 | 6 - 7 | 1225 | 68 | |
GenBank mRNA | AB064521.1 | - | 4 - 5 | 383 | 68 |
AB093293.1 | - | 5 - 6 | 1019 | 68 | |
AK013092.1 | - | 4 - 5 | 446 | 68 | |
AK029986.1 | - | 7 - 8 | 1406 | 68 | |
BC011420.1 | - | 4 - 5 | 398 | 68 | |
BC042515.1 | - | 6 - 7 | 1201 | 68 | |
BC086316.1 | - | 6 - 7 | 1293 | 68 | |
BC096446.1 | - | 6 - 7 | 1217 | 68 | |
BQ745653.1 | - | 4 - 5 | 525 | 68 |