The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010549.3 | NP_034679.1 | 1 - 2 | 112 | 97 |
GenBank mRNA | BC057664.1 | - | 1 - 2 | 42 | 97 |
BY149476.1 | - | 1 - 2 | 114 | 97 |