The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177795.1 | NP_001171266.1 | 5 - 6 | 1037 | 149 |
NM_001290372.1 | NP_001277301.1 | 4 - 5 | 620 | 149 | |
NM_021374.5 | NP_067349.2 | 4 - 5 | 690 | 149 | |
GenBank mRNA | AF191552.1 | - | 4 - 5 | 580 | 149 |
AF191554.1 | - | 4 - 5 | 566 | 149 | |
AK013773.1 | - | 4 - 5 | 519 | 149 | |
AK078208.1 | - | 4 - 5 | 535 | 149 | |
AK089947.1 | - | 4 - 5 | 510 | 149 | |
AK158864.1 | - | 4 - 5 | 531 | 149 | |
BC116925.1 | - | 4 - 5 | 580 | 149 | |
BC116929.1 | - | 4 - 5 | 580 | 149 |