Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Snrpd1
Entrez Gene ID
20641
Gene Name
small nuclear ribonucleoprotein D1
Gene Aliases
AA407109, AL023031, SMD1
Chromosome Location
Chr.18: 10617796 - 10628230 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009226.4 NP_033252.1 1 - 2 131 108
GenBank mRNA AK005517.1 - 1 - 2 121 108
AK007674.1 - 1 - 2 132 108
AK011455.1 - 1 - 2 125 108
AK013075.1 - 1 - 2 98 108
AK017788.1 - - 62 108
AK134582.1 - 1 - 2 125 108
AK135979.1 - 1 - 2 121 108
BC024875.1 - 1 - 2 117 108
M58558.1 - 1 - 2 60 108

View Full Product Details