Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Nap1l1
- Entrez Gene ID
-
53605
- Gene Name
- nucleosome assembly protein 1-like 1
- Gene Aliases
-
AA407126, AI256722, D10Ertd68e, NAP-1
- Chromosome Location
-
Chr.10: 111473192 - 111498150 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_015781.4 |
NP_056596.1 |
1 - 2 |
162 |
168 |
View Full Product Details