The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146707.1 | NP_001140179.1 | 8 - 9 | 1453 | 113 |
NM_015781.4 | NP_056596.1 | 9 - 10 | 950 | 113 | |
GenBank mRNA | AK007322.1 | - | 9 - 10 | 915 | 113 |
AK032896.1 | - | 9 - 10 | 923 | 113 | |
AK050375.1 | - | 8 - 9 | 1455 | 113 | |
AK136161.1 | - | 9 - 10 | 916 | 113 | |
AK145766.1 | - | 9 - 10 | 914 | 113 | |
AK152011.1 | - | 9 - 10 | 844 | 113 | |
AK153531.1 | - | 9 - 10 | 844 | 113 | |
AK169301.1 | - | 9 - 10 | 862 | 113 | |
AK216281.1 | - | 1 - 2 | 65 | 113 | |
BC076591.1 | - | 8 - 9 | 1001 | 113 | |
D12618.1 | - | 9 - 10 | 906 | 113 | |
X61449.1 | - | 6 - 7 | 623 | 113 |