The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011830.3 | NP_035960.2 | 1 - 2 | 130 | 77 |
GenBank mRNA | AK143505.1 | - | 1 - 2 | 171 | 77 |
AK146362.1 | - | 1 - 2 | 196 | 77 | |
AK150713.1 | - | 1 - 2 | 166 | 77 | |
AK151233.1 | - | 1 - 2 | 166 | 77 | |
AK151706.1 | - | 1 - 2 | 189 | 77 | |
AK152691.1 | - | 1 - 2 | 163 | 77 | |
AK160569.1 | - | 1 - 2 | 163 | 77 | |
AK169129.1 | - | 1 - 2 | 196 | 77 | |
AK169788.1 | - | 1 - 2 | 196 | 77 | |
BC010314.1 | - | 1 - 2 | 130 | 77 | |
BC052671.1 | - | 1 - 2 | 151 | 77 | |
M33934.1 | - | 1 - 2 | 163 | 77 | |
M98333.1 | - | 1 - 2 | 97 | 77 |