The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175331.3 | NP_780540.2 | 4 - 5 | 624 | 112 |
GenBank mRNA | AK147488.1 | - | 4 - 5 | 593 | 112 |