Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nlrx1
Entrez Gene ID
270151
Gene Name
NLR family member X1
Gene Aliases
BC034204, NOD9
Chromosome Location
Chr.9: 44252713 - 44268599 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163742.1 NP_001157214.1 2 - 3 218 65
NM_001163743.1 NP_001157215.1 2 - 3 319 65
NM_178420.3 NP_848507.2 2 - 3 307 65
GenBank mRNA AK089260.1 - 2 - 3 311 65
AK141296.1 - 2 - 3 288 65
AK145780.1 - 2 - 3 328 65
AK145988.1 - 2 - 3 308 65
AK166689.1 - 2 - 3 145 65
AK170088.1 - 2 - 3 262 65
BC034204.1 - 1 - 2 101 65
BC050054.1 - 2 - 3 214 65

View Full Product Details