The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175434.3 | NP_780643.2 | 5 - 6 | 1139 | 102 |
GenBank mRNA | AK046359.1 | - | 5 - 6 | 1119 | 102 |
AK083105.1 | - | 4 - 5 | 894 | 102 | |
BC115859.1 | - | 5 - 6 | 985 | 102 | |
BC115965.1 | - | 5 - 6 | 985 | 102 |