The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081379.2 | NP_001074848.2 | 11 - 12 | 8114 | 70 |
NR_037865.1 | 12 - 13 | 8211 | 70 | ||
GenBank mRNA | BC059876.1 | - | 3 - 4 | 2027 | 70 |
BC158043.1 | - | 10 - 11 | 7779 | 70 | |
BC172141.1 | - | 11 - 12 | 7876 | 70 |