Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Csnk2a2
Entrez Gene ID
13000
Gene Name
casein kinase 2, alpha prime polypeptide
Gene Aliases
1110035J23Rik, C77789, CK2
Chromosome Location
Chr.8: 95446096 - 95488904 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009974.3 NP_034104.1 9 - 10 1198 92
GenBank mRNA AF012251.1 - 9 - 10 923 92
AJ001420.1 - 9 - 10 1168 92
AK002927.1 - 9 - 10 1206 92
AK013008.1 - 3 - 4 238 92
AK019507.1 - 2 - 3 136 92
AK030035.1 - 3 - 4 238 92
AK076042.1 - 9 - 10 1159 92
BC047143.1 - 9 - 10 1188 92
BC057862.1 - 9 - 10 858 92

View Full Product Details