The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010332.2 | NP_034462.1 | 7 - 8 | 1555 | 114 |
GenBank mRNA | AK043210.1 | - | 7 - 8 | 1452 | 114 |
AK135555.1 | - | 7 - 8 | 1449 | 114 | |
BC008277.1 | - | 7 - 8 | 1539 | 114 |