The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016896.3 | NP_058592.1 | 11 - 12 | 2066 | 96 |
GenBank mRNA | AF143094.1 | - | 10 - 11 | 1983 | 96 |
AK036200.1 | - | 11 - 12 | 2067 | 96 | |
AK172518.1 | - | 11 - 12 | 2036 | 96 |