Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Coro6
Entrez Gene ID
216961
Gene Name
coronin 6
Gene Aliases
-
Chromosome Location
Chr.11: 77462321 - 77470484 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139130.2 NP_624356.1 4 - 5 631 69
GenBank mRNA AB070895.1 - 4 - 5 631 69
BC145575.1 - 4 - 5 708 69

View Full Product Details