Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc9a8
Entrez Gene ID
77031
Gene Name
solute carrier family 9 (sodium/hydrogen exchanger), member 8
Gene Aliases
1200006P13Rik, 6430709P13Rik, AI182282, NHE-8, NHE8
Chromosome Location
Chr.2: 167420056 - 167477000 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB089793.1 - 15 - 16 1653 67
AF482993.1 - 14 - 15 1617 67
AK049410.1 - 15 - 16 1649 67
AK051791.1 - 14 - 15 1556 67
AK129013.1 - 6 - 7 870 67
AK160826.1 - 12 - 13 1379 67
AK173069.1 - 15 - 16 1989 67
BC030879.1 - 15 - 16 1652 67
BC034508.1 - 8 - 9 1053 67
BC058947.1 - 15 - 16 1623 67

View Full Product Details