The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024927.2 | NP_001020098.1 | 13 - 14 | 1985 | 60 |
NM_001081641.1 | NP_001075110.1 | 12 - 13 | 1937 | 60 | |
GenBank mRNA | AK147444.1 | - | 13 - 14 | 1942 | 60 |
AK147630.1 | - | 12 - 13 | 1770 | 60 |