The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177449.3 | NP_803232.2 | 4 - 5 | 1313 | 56 |
GenBank mRNA | AK041606.1 | - | 5 - 6 | 1816 | 56 |
AK042435.1 | - | 5 - 6 | 1817 | 56 | |
AK148120.1 | - | 4 - 5 | 1315 | 56 |