Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lyl1
Entrez Gene ID
17095
Gene Name
lymphoblastomic leukemia 1
Gene Aliases
Lyl-1, bHLHa18
Chromosome Location
Chr.8: 84701422 - 84704716 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008535.2 NP_032561.2 2 - 3 881 67
GenBank mRNA AK076114.1 - 2 - 3 535 67
BC005736.1 - 2 - 3 501 67
CT010167.1 - 1 - 2 326 67
X57687.1 - 2 - 3 791 67

View Full Product Details