The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027840.3 | NP_082116.1 | 1 - 2 | 143 | 65 |
GenBank mRNA | AK042068.1 | - | 1 - 2 | 118 | 65 |
AK157032.1 | - | 1 - 2 | 116 | 65 | |
AK157884.1 | - | 1 - 2 | 117 | 65 | |
BC039809.1 | - | 1 - 2 | 49 | 65 |