The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301351.1 | NP_001288280.1 | 13 - 14 | 1610 | 66 |
NM_181541.4 | NP_853519.2 | 14 - 15 | 2267 | 66 | |
GenBank mRNA | AK129460.1 | - | 13 - 14 | 1523 | 66 |
AK140744.1 | - | 14 - 15 | 2270 | 66 | |
BC025636.1 | - | 14 - 15 | 2223 | 66 | |
BC125373.1 | - | 14 - 15 | 2156 | 66 | |
BC125375.1 | - | 14 - 15 | 2156 | 66 |