Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bcat1
Entrez Gene ID
12035
Gene Name
branched chain aminotransferase 1, cytosolic
Gene Aliases
BCATc, Eca39
Chromosome Location
Chr.6: 144993835 - 145076157 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024468.3 NP_001019639.1 3 - 4 519 69
NM_007532.5 NP_031558.3 3 - 4 538 69
GenBank mRNA AK012821.1 - 3 - 4 314 69
AK013888.1 - 3 - 4 475 69
AK036309.1 - 3 - 4 520 69
AK138468.1 - 3 - 4 432 69
AK166971.1 - 3 - 4 474 69
AK167232.1 - 3 - 4 465 69
AK167368.1 - 3 - 4 384 69
AK168249.1 - 3 - 4 568 69
BC053706.1 - 3 - 4 470 69
U42443.1 - 2 - 3 275 69
X17502.1 - 3 - 4 462 69

View Full Product Details