The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024468.3 | NP_001019639.1 | 8 - 9 | 78 | |
NM_007532.5 | NP_031558.3 | 8 - 9 | 78 | ||
GenBank mRNA | AK012821.1 | - | 8 - 9 | 78 | |
AK013888.1 | - | 8 - 9 | 78 | ||
AK036309.1 | - | 8 - 9 | 78 | ||
AK138468.1 | - | 8 - 9 | 78 | ||
AK166971.1 | - | 8 - 9 | 78 | ||
AK167232.1 | - | 8 - 9 | 78 | ||
AK167368.1 | - | 8 - 9 | 78 | ||
AK168249.1 | - | 8 - 9 | 1191 | 78 | |
BC053706.1 | - | 8 - 9 | 78 | ||
U42443.1 | - | 7 - 8 | 898 | 78 | |
X17502.1 | - | 8 - 9 | 1084 | 78 |