The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
GenBank mRNA | AK014418.1 | - | 6 - 7 | 1092 | 85 |
AK157860.1 | - | 6 - 7 | 958 | 85 | |
AK167113.1 | - | 6 - 7 | 1007 | 85 | |
AK178167.1 | - | 2 - 3 | 217 | 85 | |
AK207454.1 | - | 2 - 3 | 238 | 85 | |
BC005715.1 | - | 7 - 8 | 1227 | 85 | |
BC068152.1 | - | 6 - 7 | 1333 | 85 | |
D50461.1 | - | 6 - 7 | 1336 | 85 | |
U45977.1 | - | 6 - 7 | 1241 | 85 | |
U45978.1 | - | 6 - 7 | 1109 | 85 |