The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081224.2 | NP_001074693.1 | 1 - 2 | 186 | 74 |
GenBank mRNA | AK042678.1 | - | - | 347 | 74 |
AK046456.1 | - | 1 - 2 | 351 | 74 | |
AK081911.1 | - | - | 162 | 74 | |
BC132481.1 | - | 1 - 2 | 186 | 74 |