Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mogat2
Entrez Gene ID
233549
Gene Name
monoacylglycerol O-acyltransferase 2
Gene Aliases
DGAT2L5, MGAT2, Mgat1l
Chromosome Location
Chr.7: 99219084 - 99238611 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177448.4 NP_803231.1 4 - 5 695 121
GenBank mRNA AK049560.1 - 4 - 5 659 121
AK133854.1 - 4 - 5 703 121
AY157609.1 - 4 - 5 652 121
BC052831.1 - 4 - 5 699 121

View Full Product Details