Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rad51ap2
Entrez Gene ID
209550
Gene Name
RAD51 associated protein 2
Gene Aliases
EG209550
Chromosome Location
Chr.12: 11455759 - 11517161 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111118.1 NP_001104588.1 1 - 2 2731 117

View Full Product Details