Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atg16l2
Entrez Gene ID
73683
Gene Name
autophagy related 16-like 2 (S. cerevisiae)
Gene Aliases
2410118P20Rik
Chromosome Location
Chr.7: 101286313 - 101302266 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111111.1 NP_001104581.1 14 - 15 1544 91
GenBank mRNA AB476647.1 - 13 - 14 1420 91
AB476648.1 - 14 - 15 1483 91
AK010773.1 - 13 - 14 1465 91
AK131109.1 - - 1478 91
AK155125.1 - 14 - 15 1534 91
AK157410.1 - 14 - 15 1523 92
AK158186.1 - 13 - 14 1658 91
BC060054.1 - 13 - 14 2045 91

View Full Product Details