The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001162924.1 | NP_001156396.1 | 1 - 2 | 209 | 84 |
GenBank mRNA | BC090668.1 | - | 1 - 2 | 209 | 84 |